The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000705.3 | NP_000696.1 | 3 - 4 | 407 | 82 |
GenBank mRNA | AK310129.1 | - | 3 - 4 | 82 | |
BC029059.1 | - | 3 - 4 | 395 | 82 | |
DA964061.1 | - | 3 - 4 | 407 | 82 | |
M75110.1 | - | 3 - 4 | 370 | 82 |