The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000705.3 | NP_000696.1 | 5 - 6 | 668 | 110 |
GenBank mRNA | BC029059.1 | - | 5 - 6 | 656 | 110 |
M75110.1 | - | 5 - 6 | 631 | 110 |