The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001193343.1 | NP_001180272.1 | 3 - 4 | 291 | 71 |
NM_018135.3 | NP_060605.1 | 3 - 4 | 291 | 71 | |
XM_006715134.2 | XP_006715197.1 | 3 - 4 | 267 | 71 | |
GenBank mRNA | AB049952.1 | - | 3 - 4 | 268 | 71 |
AK001410.1 | - | 3 - 4 | 297 | 71 | |
AK300757.1 | - | 3 - 4 | 262 | 71 | |
BC010364.1 | - | 3 - 4 | 266 | 71 | |
DQ884400.1 | - | 3 - 4 | 258 | 71 |