The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018135.3 | NP_060605.1 | 4 - 5 | 411 | 65 |
GenBank mRNA | AB049952.1 | - | 4 - 5 | 388 | 65 |
AK001410.1 | - | 4 - 5 | 417 | 65 | |
BC010364.1 | - | 4 - 5 | 386 | 65 |