The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018323.3 | NP_060793.2 | 3 - 4 | 760 | 84 |
XM_005248174.1 | XP_005248231.1 | 3 - 4 | 758 | 84 | |
XM_005248175.4 | XP_005248232.1 | 3 - 4 | 538 | 84 | |
GenBank mRNA | AF411320.1 | - | 3 - 4 | 627 | 84 |
AK001967.1 | - | 3 - 4 | 748 | 84 | |
AK128622.1 | - | 4 - 5 | 665 | 84 | |
AK296302.1 | - | 3 - 4 | 450 | 84 | |
AY065990.1 | - | 3 - 4 | 738 | 84 | |
AY091514.1 | - | 3 - 4 | 747 | 84 | |
BC051749.1 | - | 3 - 4 | 760 | 84 |