Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PPP2R1A
Entrez Gene ID
5518
Gene Name
protein phosphatase 2 scaffold subunit Aalpha
Gene Aliases
MRD36, PP2A-Aalpha, PP2AAALPHA, PR65A
Chromosome Location
Chr.19: 52189802 - 52226425 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014225.5 NP_055040.2 12 - 13 1815 60
NR_033500.1 11 - 12 1714 60
XM_017026929.1 XP_016882418.1 12 - 13 2044 60
GenBank mRNA AK090488.1 - 12 - 13 2049 60
AK091387.1 - 12 - 13 1419 60
AK130000.1 - 11 - 12 1323 60
AK290593.1 - 12 - 13 1603 60
AK291932.1 - 12 - 13 1598 60
AK293174.1 - 13 - 14 1566 60
AK293493.1 - 11 - 12 1439 60
AK299012.1 - 12 - 13 1470 60
AK303934.1 - - 271 60
AK315700.1 - 12 - 13 1631 60
BC001537.2 - 12 - 13 1561 60
BC026733.1 - 12 - 13 1578 60
CR450340.1 - 12 - 13 1520 60
J02902.1 - 12 - 13 1664 60

View Full Product Details