The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014225.5 | NP_055040.2 | 4 - 5 | 797 | 96 |
NR_033500.1 | 3 - 4 | 696 | 96 | ||
XM_017026929.1 | XP_016882418.1 | 4 - 5 | 1026 | 96 | |
GenBank mRNA | AK090488.1 | - | 4 - 5 | 1031 | 96 |
AK091387.1 | - | 4 - 5 | 401 | 96 | |
AK290593.1 | - | 4 - 5 | 585 | 96 | |
AK291932.1 | - | 4 - 5 | 580 | 96 | |
AK293174.1 | - | 5 - 6 | 548 | 96 | |
AK293493.1 | - | 3 - 4 | 421 | 96 | |
AK299012.1 | - | 4 - 5 | 452 | 96 | |
AK303934.1 | - | - | 898 | 96 | |
AK307743.1 | - | 4 - 5 | 668 | 96 | |
AK315700.1 | - | 4 - 5 | 613 | 96 | |
BC001537.2 | - | 4 - 5 | 543 | 96 | |
BC026733.1 | - | 4 - 5 | 560 | 96 | |
CR450340.1 | - | 4 - 5 | 502 | 96 | |
J02902.1 | - | 4 - 5 | 646 | 96 |