The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001238.3 | NP_001229.1 | 9 - 10 | 1022 | 68 |
NM_001322261.1 | NP_001309190.1 | 8 - 9 | 875 | 68 | |
NM_001322262.1 | NP_001309191.1 | 8 - 9 | 975 | 68 | |
XM_011527440.2 | XP_011525742.1 | 9 - 10 | 908 | 68 | |
GenBank mRNA | AK291549.1 | - | 9 - 10 | 1022 | 68 |
BC035498.2 | - | 9 - 10 | 1012 | 68 | |
KF672848.1 | - | 8 - 9 | 875 | 68 | |
M73812.1 | - | 7 - 8 | 851 | 68 | |
M74093.1 | - | 7 - 8 | 819 | 68 |