Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ARHGEF5
Entrez Gene ID
7984
Gene Name
Rho guanine nucleotide exchange factor 5
Gene Aliases
GEF5, P60, TIM, TIM1
Chromosome Location
Chr.7: 144355341 - 144380632 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005435.3 NP_005426.2 10 - 11 4255 66
GenBank mRNA AK090570.1 - 10 - 11 1867 66
AK123596.1 - 9 - 10 2454 66
AK160365.1 - 10 - 11 4311 66
BC010046.2 - 10 - 11 1200 66
BC011839.2 - 10 - 11 1200 66
BC014555.2 - 10 - 11 1200 66
BC136661.1 - 10 - 11 4169 66
BX649178.1 - 9 - 10 2140 66
CR749203.1 - 10 - 11 4195 66
U02082.1 - 8 - 9 996 66

Gene Symbol
ARHGEF34P
Entrez Gene ID
728377
Gene Name
Rho guanine nucleotide exchange factor 34, pseudogene
Gene Aliases
-
Chromosome Location
Chr.7: 144258996 - 144286003 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_033942.1 9 - 10 3173 66
GenBank mRNA BC107871.1 - 3 - 4 194 66
DQ786236.1 - 4 - 5 310 66

View Full Product Details