Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ARHGEF5
- Entrez Gene ID
-
7984
- Gene Name
- Rho guanine nucleotide exchange factor 5
- Gene Aliases
-
GEF5, P60, TIM, TIM1
- Chromosome Location
-
Chr.7: 144355341 - 144380632 on Build GRCh38
- Gene Symbol
- ARHGEF34P
- Entrez Gene ID
-
728377
- Gene Name
- Rho guanine nucleotide exchange factor 34, pseudogene
- Gene Aliases
-
-
- Chromosome Location
-
Chr.7: 144258996 - 144286003 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_033942.1 |
|
8 - 9 |
3085 |
76 |
GenBank mRNA |
BC107871.1 |
- |
2 - 3 |
106 |
76 |
DQ786236.1 |
- |
3 - 4 |
222 |
76 |
View Full Product Details