The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021161.4 | NP_066984.1 | 5 - 6 | 1309 | 96 |
NM_138317.2 | NP_612190.1 | 5 - 6 | 1318 | 96 | |
NM_138318.2 | NP_612191.1 | 5 - 6 | 989 | 96 | |
XM_011536840.2 | XP_011535142.1 | 4 - 5 | 694 | 96 | |
GenBank mRNA | AF279890.1 | - | 5 - 6 | 1324 | 96 |
AF385399.1 | - | 5 - 6 | 990 | 96 | |
AF385400.1 | - | 5 - 6 | 1072 | 96 | |
AK290518.1 | - | 5 - 6 | 96 | ||
AK313499.1 | - | 5 - 6 | 1318 | 96 | |
AK315263.1 | - | 5 - 6 | 977 | 96 | |
BC075021.2 | - | 5 - 6 | 908 | 96 | |
BC075022.2 | - | 5 - 6 | 905 | 96 | |
EU978938.1 | - | 5 - 6 | 1138 | 96 | |
EU978939.1 | - | 5 - 6 | 991 | 96 | |
EU978940.1 | - | 5 - 6 | 895 | 96 | |
EU978941.1 | - | 5 - 6 | 895 | 96 |