Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LY96
Entrez Gene ID
23643
Gene Name
lymphocyte antigen 96
Gene Aliases
ESOP-1, MD-2, MD2, ly-96
Chromosome Location
Chr.8: 73991275 - 74099807 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001195797.1 NP_001182726.1 3 - 4 413 101
NM_015364.4 NP_056179.3 4 - 5 503 101
GenBank mRNA AB018549.1 - 4 - 5 514 101
AB446498.1 - 4 - 5 389 101
AF168121.1 - 4 - 5 410 101
BC020690.1 - 4 - 5 413 101

View Full Product Details