The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014663.2 | NP_055478.2 | 19 - 20 | 3009 | 73 |
XM_005271354.3 | XP_005271411.1 | 19 - 20 | 3046 | 73 | |
XM_005271355.3 | XP_005271412.1 | 19 - 20 | 2964 | 73 | |
XM_005271356.3 | XP_005271413.1 | 20 - 21 | 3085 | 73 | |
XM_011542467.2 | XP_011540769.2 | 10 - 11 | 2117 | 73 | |
XM_017002912.1 | XP_016858401.1 | 20 - 21 | 3154 | 73 | |
GenBank mRNA | AB014577.1 | - | 19 - 20 | 2968 | 73 |
AK294596.1 | - | 5 - 6 | 594 | 73 | |
BC002558.2 | - | 19 - 20 | 2977 | 73 |