The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004863.3 | NP_004854.1 | 6 - 7 | 1042 | 94 |
XM_011537384.2 | XP_011535686.1 | 6 - 7 | 938 | 94 | |
GenBank mRNA | AB011098.1 | - | 6 - 7 | 1042 | 94 |
BC005123.2 | - | 6 - 7 | 943 | 94 | |
BQ338973.1 | - | 3 - 4 | 374 | 94 | |
U15555.1 | - | 5 - 6 | 678 | 94 | |
Y08686.1 | - | 6 - 7 | 902 | 94 |