The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020754.3 | NP_065805.2 | 2 - 3 | 737 | 83 |
XM_006713714.3 | XP_006713777.1 | 2 - 3 | 821 | 83 | |
GenBank mRNA | AB033030.1 | - | 2 - 3 | 737 | 83 |
AK293726.1 | - | 2 - 3 | 334 | 83 | |
BC112163.1 | - | 2 - 3 | 232 | 83 | |
BC112165.1 | - | 2 - 3 | 232 | 83 |