Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRIM31
Entrez Gene ID
11074
Gene Name
tripartite motif containing 31
Gene Aliases
C6orf13, HCG1, HCGI, RNF
Chromosome Location
Chr.6: 30102892 - 30113106 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007028.4 NP_008959.3 4 - 5 870 109
NR_134870.1 4 - 5 870 109
XM_011514265.1 XP_011512567.1 4 - 5 854 109
GenBank mRNA AF230386.1 - 3 - 4 853 109
AK222729.1 - 4 - 5 854 109
AK290270.1 - 4 - 5 857 109
BC016866.1 - 1 - 2 103 109
BC017017.1 - 3 - 4 831 109
BT006675.1 - 3 - 4 744 109

View Full Product Details