The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000541.4 | NP_000532.2 | 1 - 2 | 355 | 59 |
GenBank mRNA | AA317634.1 | - | 1 - 2 | 181 | 59 |
BQ639460.1 | - | 1 - 2 | 168 | 59 | |
BX641043.1 | - | 1 - 2 | 86 | 59 | |
BX647821.1 | - | 1 - 2 | 139 | 59 | |
BX647827.1 | - | 1 - 2 | 87 | 59 | |
DQ980620.1 | - | 1 - 2 | 175 | 59 |