The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000541.4 | NP_000532.2 | 2 - 3 | 465 | 83 |
XM_017004641.1 | XP_016860130.1 | 1 - 2 | 154 | 83 | |
XM_017004642.1 | XP_016860131.1 | 1 - 2 | 154 | 83 | |
XM_017004643.1 | XP_016860132.1 | 1 - 2 | 154 | 83 | |
GenBank mRNA | AA317634.1 | - | 2 - 3 | 291 | 83 |
BQ639460.1 | - | 2 - 3 | 278 | 83 | |
BX491422.1 | - | 2 - 3 | 361 | 83 | |
BX647821.1 | - | 2 - 3 | 249 | 83 | |
BX647827.1 | - | 2 - 3 | 197 | 83 | |
DQ980620.1 | - | 2 - 3 | 285 | 83 | |
X12453.1 | - | 2 - 3 | 298 | 83 |