Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DTNB
Entrez Gene ID
1838
Gene Name
dystrobrevin beta
Gene Aliases
-
Chromosome Location
Chr.2: 25377220 - 25673647 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001256303.1 NP_001243232.1 5 - 6 717 144
NM_001256304.1 NP_001243233.1 4 - 5 454 144
NM_001256308.2 NP_001243237.1 4 - 5 588 144
NM_001320933.1 NP_001307862.1 5 - 6 717 144
NM_001320934.1 NP_001307863.1 5 - 6 717 144
NM_001320936.1 NP_001307865.1 5 - 6 717 144
NM_001320937.1 NP_001307866.1 5 - 6 717 144
NM_021907.4 NP_068707.1 5 - 6 717 144
NM_033147.3 NP_149159.2 5 - 6 717 144
NM_033148.3 NP_149160.1 5 - 6 717 144
NM_183360.2 NP_899204.1 5 - 6 717 144
NM_183361.2 NP_899205.1 5 - 6 717 144
XM_005264182.1 XP_005264239.1 5 - 6 634 144
XM_011532671.1 XP_011530973.1 5 - 6 634 144
XM_011532672.2 XP_011530974.1 5 - 6 634 144
XM_011532673.1 XP_011530975.1 5 - 6 634 144
XM_011532674.1 XP_011530976.1 5 - 6 634 144
XM_011532675.1 XP_011530977.1 5 - 6 634 144
XM_011532676.1 XP_011530978.1 5 - 6 634 144
XM_011532677.2 XP_011530979.1 4 - 5 1090 144
XM_011532679.1 XP_011530981.1 5 - 6 634 144
XM_017003529.1 XP_016859018.1 5 - 6 634 144
XM_017003530.1 XP_016859019.1 5 - 6 634 144
XM_017003531.1 XP_016859020.1 5 - 6 634 144
XM_017003532.1 XP_016859021.1 5 - 6 634 144
XM_017003533.1 XP_016859022.1 5 - 6 634 144
XM_017003534.1 XP_016859023.1 5 - 6 634 144
XM_017003535.1 XP_016859024.1 5 - 6 634 144
XM_017003536.1 XP_016859025.1 5 - 6 634 144
GenBank mRNA AF022728.1 - 5 - 6 615 144
AK094494.1 - 5 - 6 580 144
AK300019.1 - 5 - 6 594 144
AL555818.3 - 5 - 6 631 144
BC016655.1 - 5 - 6 610 144
BC049366.1 - 5 - 6 708 144
BT009805.1 - 4 - 5 454 144
BU846095.1 - 3 - 4 395 144
DQ160290.1 - 4 - 5 454 144
Y12712.1 - 5 - 6 637 144
Y15718.1 - 4 - 5 454 144
Y15722.1 - 4 - 5 454 144

View Full Product Details