Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DTNB
Entrez Gene ID
1838
Gene Name
dystrobrevin beta
Gene Aliases
-
Chromosome Location
Chr.2: 25377220 - 25673647 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001256303.1 NP_001243232.1 9 - 10 1258 83
NM_001256304.1 NP_001243233.1 8 - 9 995 83
NM_001256308.2 NP_001243237.1 8 - 9 1129 83
NM_001320932.1 NP_001307861.1 8 - 9 1103 83
NM_001320933.1 NP_001307862.1 9 - 10 1258 83
NM_001320934.1 NP_001307863.1 9 - 10 1258 83
NM_001320935.1 NP_001307864.1 7 - 8 1017 83
NM_001320936.1 NP_001307865.1 9 - 10 1258 83
NM_021907.4 NP_068707.1 9 - 10 1258 83
NM_033147.3 NP_149159.2 9 - 10 1258 83
NM_033148.3 NP_149160.1 9 - 10 1258 83
NM_183360.2 NP_899204.1 9 - 10 1258 83
NM_183361.2 NP_899205.1 9 - 10 1258 83
XM_005264182.1 XP_005264239.1 9 - 10 1175 83
XM_011532671.1 XP_011530973.1 9 - 10 1175 83
XM_011532672.2 XP_011530974.1 9 - 10 1175 83
XM_011532673.1 XP_011530975.1 9 - 10 1175 83
XM_011532674.1 XP_011530976.1 9 - 10 1175 83
XM_011532675.1 XP_011530977.1 9 - 10 1175 83
XM_011532676.1 XP_011530978.1 9 - 10 1175 83
XM_011532677.2 XP_011530979.1 8 - 9 1631 83
XM_011532679.1 XP_011530981.1 9 - 10 1175 83
XM_011532680.2 XP_011530982.1 8 - 9 1096 83
XM_011532681.2 XP_011530983.1 8 - 9 1089 83
XM_017003529.1 XP_016859018.1 9 - 10 1175 83
XM_017003530.1 XP_016859019.1 9 - 10 1175 83
XM_017003531.1 XP_016859020.1 9 - 10 1175 83
XM_017003532.1 XP_016859021.1 9 - 10 1175 83
XM_017003533.1 XP_016859022.1 9 - 10 1175 83
XM_017003534.1 XP_016859023.1 9 - 10 1175 83
XM_017003535.1 XP_016859024.1 9 - 10 1175 83
XM_017003536.1 XP_016859025.1 9 - 10 1175 83
XM_017003537.1 XP_016859026.1 5 - 6 1528 83
XM_017003538.1 XP_016859027.1 8 - 9 1021 83
XM_017003539.1 XP_016859028.1 8 - 9 1032 83
XM_017003540.1 XP_016859029.1 8 - 9 1089 83
XM_017003541.1 XP_016859030.1 8 - 9 1073 83
XM_017003542.1 XP_016859031.1 8 - 9 1027 83
XM_017003543.1 XP_016859032.1 7 - 8 921 83
XM_017003544.1 XP_016859033.1 8 - 9 1027 83
XM_017003545.1 XP_016859034.1 7 - 8 921 83
GenBank mRNA AF022728.1 - 9 - 10 1156 83
AF070567.1 - 3 - 4 400 83
AK225724.1 - 8 - 9 1006 83
AK294174.1 - 7 - 8 922 83
AK294357.1 - 8 - 9 939 83
AK300019.1 - 9 - 10 1135 83
AK301386.1 - 8 - 9 1131 83
BC016655.1 - 9 - 10 1151 83
BC049366.1 - 9 - 10 1249 83
BT009805.1 - 8 - 9 995 83
BX389926.1 - 3 - 4 378 83
DQ160290.1 - 8 - 9 995 83
R15062.1 - 1 - 2 108 83
Y12712.1 - 9 - 10 1178 83
Y15718.1 - 8 - 9 995 83
Y15722.1 - 8 - 9 995 83

View Full Product Details