Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZFPL1
Entrez Gene ID
7542
Gene Name
zinc finger protein like 1
Gene Aliases
D11S750, MCG4
Chromosome Location
Chr.11: 65084222 - 65088402 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006782.3 NP_006773.2 5 - 6 646 77
GenBank mRNA AF030291.1 - 5 - 6 633 77
AF043611.1 - 5 - 6 510 77
AK291964.1 - 5 - 6 627 77
BC012530.2 - 5 - 6 581 77
BC012814.2 - 5 - 6 581 77

View Full Product Details