Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KIF20B
Entrez Gene ID
9585
Gene Name
kinesin family member 20B
Gene Aliases
CT90, KRMP1, MPHOSPH1, MPP-1, MPP1
Chromosome Location
Chr.10: 89701590 - 89774943 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001284259.1 NP_001271188.1 32 - 33 5475 94
NM_016195.3 NP_057279.2 32 - 33 5355 94
GenBank mRNA AB033337.1 - 33 - 34 5428 94
AK025628.1 - 12 - 13 1510 94
AL117496.1 - 32 - 33 5310 94
AL137392.1 - 19 - 20 3942 94
AY282407.1 - 17 - 18 3272 94
AY739715.1 - 14 - 15 2374 94
L16782.1 - 13 - 14 1622 94
U93121.1 - 13 - 14 2183 94

View Full Product Details