Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HSF1
Entrez Gene ID
3297
Gene Name
heat shock transcription factor 1
Gene Aliases
HSTF1
Chromosome Location
Chr.8: 144291588 - 144314722 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005526.2 NP_005517.1 9 - 10 1310 76
XM_017013377.1 XP_016868866.1 9 - 10 1310 76
GenBank mRNA AK054762.1 - 5 - 6 1427 76
AK125467.1 - 6 - 7 2612 76
AK222497.1 - 9 - 10 1294 76
AK290975.1 - 9 - 10 1290 76
AK300205.1 - 6 - 7 792 76
BC014638.1 - 9 - 10 1281 76
BT007351.1 - 9 - 10 1140 76
M64673.1 - 9 - 10 1300 76

View Full Product Details