Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MADD
Entrez Gene ID
8567
Gene Name
MAP kinase activating death domain
Gene Aliases
DENN, IG20, RAB3GEP
Chromosome Location
Chr.11: 47269376 - 47330031 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135943.1 NP_001129415.1 27 - 28 4281 65
NM_001135944.1 NP_001129416.1 27 - 28 4248 65
NM_003682.3 NP_003673.3 30 - 31 4566 65
NM_130470.2 NP_569826.2 27 - 28 4555 65
NM_130471.2 NP_569827.2 28 - 29 4320 65
NM_130472.2 NP_569828.2 27 - 28 4260 65
NM_130473.2 NP_569829.2 28 - 29 4449 65
NM_130474.2 NP_569830.2 27 - 28 4260 65
NM_130475.2 NP_569831.1 30 - 31 4590 65
NM_130476.2 NP_569832.2 27 - 28 4410 65
XM_005253189.2 XP_005253246.1 30 - 31 4590 65
XM_005253196.2 XP_005253253.1 28 - 29 4473 65
XM_005253199.2 XP_005253256.1 27 - 28 4413 65
XM_005253200.2 XP_005253257.1 30 - 31 4587 65
XM_005253201.2 XP_005253258.1 28 - 29 4344 65
XM_005253203.2 XP_005253260.1 27 - 28 4284 65
XM_005253204.2 XP_005253261.1 27 - 28 4272 65
XM_005253205.1 XP_005253262.1 27 - 28 4284 65
XM_017018478.1 XP_016873967.1 30 - 31 4578 65
XM_017018479.1 XP_016873968.1 29 - 30 4524 65
XM_017018480.1 XP_016873969.1 28 - 29 4470 65
XM_017018481.1 XP_016873970.1 28 - 29 4464 65
XM_017018482.1 XP_016873971.1 28 - 29 4461 65
XM_017018483.1 XP_016873972.1 30 - 31 4449 65
XM_017018484.1 XP_016873973.1 27 - 28 4401 65
XM_017018485.1 XP_016873974.1 27 - 28 4362 65
XM_017018486.1 XP_016873975.1 29 - 30 4395 65
XM_017018487.1 XP_016873976.1 29 - 30 4389 65
XM_017018488.1 XP_016873977.1 30 - 31 4578 65
XM_017018489.1 XP_016873978.1 30 - 31 4539 65
XM_017018490.1 XP_016873979.1 29 - 30 4523 65
XM_017018491.1 XP_016873980.1 29 - 30 4489 65
XM_017018492.1 XP_016873981.1 29 - 30 4524 65
XM_017018493.1 XP_016873982.1 29 - 30 4505 65
XM_017018494.1 XP_016873983.1 28 - 29 4476 65
XM_017018495.1 XP_016873984.1 28 - 29 4473 65
XM_017018496.1 XP_016873985.1 28 - 29 4420 65
XM_017018497.1 XP_016873986.1 28 - 29 4425 65
XM_017018498.1 XP_016873987.1 30 - 31 4449 65
XM_017018499.1 XP_016873988.1 30 - 31 4418 65
XM_017018500.1 XP_016873989.1 27 - 28 4410 65
XM_017018501.1 XP_016873990.1 29 - 30 4363 65
XM_017018502.1 XP_016873991.1 29 - 30 4376 65
XM_017018503.1 XP_016873992.1 29 - 30 4360 65
XM_017018504.1 XP_016873993.1 29 - 30 4373 65
XM_017018505.1 XP_016873994.1 29 - 30 4351 65
XM_017018506.1 XP_016873995.1 29 - 30 4356 65
XM_017018507.1 XP_016873996.1 29 - 30 4376 65
XM_017018508.1 XP_016873997.1 28 - 29 4347 65
XM_017018509.1 XP_016873998.1 28 - 29 4300 65
XM_017018510.1 XP_016873999.1 28 - 29 4335 65
GenBank mRNA AB002356.1 - 30 - 31 4562 65
AF440100.1 - 30 - 31 4540 65
AF440101.1 - 28 - 29 4423 65
AF440102.1 - 28 - 29 4294 65
AF440103.1 - 27 - 28 4234 65
AF440434.1 - 27 - 28 4234 65
AK292442.1 - 27 - 28 4212 65
AK308780.1 - 11 - 12 1018 65
BC040484.1 - 27 - 28 4265 65
U44953.1 - 27 - 28 4374 65
U77352.1 - 27 - 28 4523 65

View Full Product Details