Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MADD
Entrez Gene ID
8567
Gene Name
MAP kinase activating death domain
Gene Aliases
DENN, IG20, RAB3GEP
Chromosome Location
Chr.11: 47269376 - 47330031 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135943.1 NP_001129415.1 9 - 10 1916 61
NM_001135944.1 NP_001129416.1 9 - 10 1892 61
NM_003682.3 NP_003673.3 9 - 10 1892 61
NM_130470.2 NP_569826.2 9 - 10 2058 61
NM_130471.2 NP_569827.2 9 - 10 1892 61
NM_130472.2 NP_569828.2 9 - 10 1892 61
NM_130473.2 NP_569829.2 9 - 10 1892 61
NM_130474.2 NP_569830.2 9 - 10 1892 61
NM_130475.2 NP_569831.1 9 - 10 1916 61
NM_130476.2 NP_569832.2 9 - 10 1916 61
XM_005253189.2 XP_005253246.1 9 - 10 1916 61
XM_005253196.2 XP_005253253.1 9 - 10 1916 61
XM_005253199.2 XP_005253256.1 9 - 10 1916 61
XM_005253200.2 XP_005253257.1 9 - 10 1916 61
XM_005253201.2 XP_005253258.1 9 - 10 1916 61
XM_005253203.2 XP_005253260.1 9 - 10 1916 61
XM_005253204.2 XP_005253261.1 9 - 10 1916 61
XM_005253205.1 XP_005253262.1 9 - 10 1916 61
XM_017018478.1 XP_016873967.1 9 - 10 1916 61
XM_017018479.1 XP_016873968.1 9 - 10 1916 61
XM_017018480.1 XP_016873969.1 9 - 10 1916 61
XM_017018481.1 XP_016873970.1 9 - 10 1916 61
XM_017018482.1 XP_016873971.1 9 - 10 1916 61
XM_017018483.1 XP_016873972.1 9 - 10 1916 61
XM_017018484.1 XP_016873973.1 9 - 10 1916 61
XM_017018485.1 XP_016873974.1 9 - 10 1877 61
XM_017018486.1 XP_016873975.1 9 - 10 1916 61
XM_017018487.1 XP_016873976.1 9 - 10 1916 61
XM_017018488.1 XP_016873977.1 9 - 10 1916 61
XM_017018489.1 XP_016873978.1 9 - 10 1877 61
XM_017018490.1 XP_016873979.1 9 - 10 1903 61
XM_017018491.1 XP_016873980.1 9 - 10 1872 61
XM_017018492.1 XP_016873981.1 9 - 10 1916 61
XM_017018493.1 XP_016873982.1 9 - 10 1903 61
XM_017018494.1 XP_016873983.1 9 - 10 1916 61
XM_017018495.1 XP_016873984.1 9 - 10 1916 61
XM_017018496.1 XP_016873985.1 9 - 10 1872 61
XM_017018497.1 XP_016873986.1 9 - 10 1877 61
XM_017018498.1 XP_016873987.1 9 - 10 1916 61
XM_017018499.1 XP_016873988.1 9 - 10 1885 61
XM_017018500.1 XP_016873989.1 9 - 10 1916 61
XM_017018501.1 XP_016873990.1 9 - 10 1872 61
XM_017018502.1 XP_016873991.1 9 - 10 1885 61
XM_017018503.1 XP_016873992.1 9 - 10 1872 61
XM_017018504.1 XP_016873993.1 9 - 10 1885 61
XM_017018505.1 XP_016873994.1 9 - 10 1872 61
XM_017018506.1 XP_016873995.1 9 - 10 1877 61
XM_017018507.1 XP_016873996.1 9 - 10 1903 61
XM_017018508.1 XP_016873997.1 9 - 10 1916 61
XM_017018509.1 XP_016873998.1 9 - 10 1872 61
XM_017018510.1 XP_016873999.1 9 - 10 1916 61
GenBank mRNA AB002356.1 - 9 - 10 1888 61
AF440100.1 - 9 - 10 1866 61
AF440101.1 - 9 - 10 1866 61
AF440102.1 - 9 - 10 1866 61
AF440103.1 - 9 - 10 1866 61
AF440434.1 - 9 - 10 1866 61
AK292442.1 - 9 - 10 1856 61
BC040484.1 - 9 - 10 1900 61
U44953.1 - 9 - 10 1880 61
U77352.1 - 9 - 10 2026 61

View Full Product Details