Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HYAL4
Entrez Gene ID
23553
Gene Name
hyaluronoglucosaminidase 4
Gene Aliases
CSHY
Chromosome Location
Chr.7: 123763708 - 123877481 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012269.2 NP_036401.2 3 - 4 1589 77
XM_011515990.2 XP_011514292.1 3 - 4 1665 77
XM_017011911.1 XP_016867400.1 4 - 5 1292 77
GenBank mRNA AB470346.1 - 2 - 3 1021 77
AF009010.1 - 3 - 4 1592 77
BC104788.1 - 2 - 3 1027 77
BC104790.1 - 2 - 3 1027 77

View Full Product Details