The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001272020.1 | NP_001258949.1 | 3 - 4 | 1561 | 63 |
NM_004796.5 | NP_004787.2 | 14 - 15 | 3014 | 63 | |
NM_138970.4 | NP_620426.2 | 3 - 4 | 1561 | 63 | |
XM_011537367.1 | XP_011535669.1 | 19 - 20 | 4664 | 63 | |
XM_011537373.1 | XP_011535675.1 | 19 - 20 | 4664 | 63 | |
XM_017021799.1 | XP_016877288.1 | 19 - 20 | 4664 | 63 | |
XM_017021800.1 | XP_016877289.1 | 19 - 20 | 4664 | 63 | |
XM_017021801.1 | XP_016877290.1 | 18 - 19 | 4637 | 63 | |
XM_017021802.1 | XP_016877291.1 | 17 - 18 | 4658 | 63 | |
XM_017021807.1 | XP_016877296.1 | 19 - 20 | 4664 | 63 | |
GenBank mRNA | AB018286.1 | - | 14 - 15 | 2995 | 63 |
AK056530.1 | - | 3 - 4 | 1561 | 63 | |
AL833739.1 | - | 3 - 4 | 856 | 63 | |
BC059368.1 | - | 3 - 4 | 1487 | 63 | |
BC068469.1 | - | 3 - 4 | 1487 | 63 | |
BC152457.1 | - | 14 - 15 | 2995 | 63 |