Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NRXN3
Entrez Gene ID
9369
Gene Name
neurexin 3
Gene Aliases
C14orf60
Chromosome Location
Chr.14: 77979904 - 79868290 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004796.5 NP_004787.2 3 - 4 602 72
NR_073547.1 6 - 7 2210 72
XM_005268218.2 XP_005268275.1 8 - 9 2252 72
XM_006720322.2 XP_006720385.1 7 - 8 2240 72
XM_006720323.2 XP_006720386.1 6 - 7 2216 72
XM_011537363.1 XP_011535665.1 8 - 9 2252 72
XM_011537364.2 XP_011535666.1 9 - 10 2214 72
XM_011537365.2 XP_011535667.1 8 - 9 2040 72
XM_011537366.1 XP_011535668.1 7 - 8 2228 72
XM_011537367.1 XP_011535669.1 8 - 9 2252 72
XM_011537368.1 XP_011535670.1 8 - 9 2252 72
XM_011537369.1 XP_011535671.1 8 - 9 2252 72
XM_011537370.1 XP_011535672.1 7 - 8 2228 72
XM_011537371.1 XP_011535673.1 8 - 9 2252 72
XM_011537372.1 XP_011535674.1 8 - 9 2252 72
XM_011537373.1 XP_011535675.1 8 - 9 2252 72
XM_011537377.2 XP_011535679.1 4 - 5 544 72
XM_017021790.1 XP_016877279.1 9 - 10 2352 72
XM_017021791.1 XP_016877280.1 9 - 10 2135 72
XM_017021792.1 XP_016877281.1 7 - 8 2035 72
XM_017021793.1 XP_016877282.1 8 - 9 2252 72
XM_017021794.1 XP_016877283.1 6 - 7 2216 72
XM_017021795.1 XP_016877284.1 6 - 7 2011 72
XM_017021796.1 XP_016877285.1 7 - 8 2228 72
XM_017021797.1 XP_016877286.1 5 - 6 2192 72
XM_017021798.1 XP_016877287.1 6 - 7 2216 72
XM_017021799.1 XP_016877288.1 8 - 9 2252 72
XM_017021800.1 XP_016877289.1 8 - 9 2252 72
XM_017021801.1 XP_016877290.1 8 - 9 2252 72
XM_017021802.1 XP_016877291.1 6 - 7 2246 72
XM_017021803.1 XP_016877292.1 7 - 8 2231 72
XM_017021804.1 XP_016877293.1 8 - 9 2252 72
XM_017021805.1 XP_016877294.1 6 - 7 2216 72
XM_017021806.1 XP_016877295.1 7 - 8 2228 72
XM_017021807.1 XP_016877296.1 8 - 9 2252 72
GenBank mRNA AB018286.1 - 3 - 4 583 72
AJ316284.2 - 6 - 7 1998 72
AK310235.1 - 3 - 4 555 72
BC152457.1 - 3 - 4 583 72

View Full Product Details