The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_052971.2 | NP_443203.1 | 2 - 3 | 223 | 69 |
GenBank mRNA | AJ306405.1 | - | 2 - 3 | 223 | 69 |
AJ409065.1 | - | 2 - 3 | 1567 | 69 | |
BC070199.1 | - | 2 - 3 | 221 | 69 | |
BX443829.2 | - | 2 - 3 | 211 | 69 |