The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001017405.2 | NP_001017405.1 | 6 - 7 | 825 | 79 |
NM_001297430.1 | NP_001284359.1 | 5 - 6 | 621 | 79 | |
NM_001297431.1 | NP_001284360.1 | 4 - 5 | 625 | 79 | |
NM_001297432.1 | NP_001284361.1 | 6 - 7 | 963 | 79 | |
NM_001297433.1 | NP_001284362.1 | 6 - 7 | 899 | 79 | |
NM_005882.4 | NP_005873.2 | 5 - 6 | 702 | 79 | |
NR_123716.1 | 4 - 5 | 625 | 79 | ||
XM_006713849.3 | XP_006713912.1 | 6 - 7 | 785 | 79 | |
XM_006713850.2 | XP_006713913.1 | 6 - 7 | 958 | 79 | |
XM_011513376.1 | XP_011511678.1 | 7 - 8 | 945 | 79 | |
XM_011513377.1 | XP_011511679.1 | 6 - 7 | 863 | 79 | |
XM_017007636.1 | XP_016863125.1 | 7 - 8 | 945 | 79 | |
XM_017007637.1 | XP_016863126.1 | 6 - 7 | 862 | 79 | |
GenBank mRNA | AB208907.1 | - | 1 - 2 | 3525 | 79 |
AK001088.1 | - | 6 - 7 | 785 | 79 | |
AK022515.1 | - | 4 - 5 | 594 | 79 | |
AK022586.1 | - | 5 - 6 | 662 | 79 | |
AK091951.1 | - | 1 - 2 | 696 | 79 | |
AK127875.1 | - | 2 - 3 | 1053 | 79 | |
AK128302.1 | - | 1 - 2 | 302 | 79 | |
AK296174.1 | - | 5 - 6 | 578 | 79 | |
AK298944.1 | - | 6 - 7 | 879 | 79 | |
AK300572.1 | - | 7 - 8 | 722 | 79 | |
AK301155.1 | - | 6 - 7 | 793 | 79 | |
AY189687.1 | - | 6 - 7 | 738 | 79 | |
AY236486.1 | - | 4 - 5 | 570 | 79 | |
BC001225.1 | - | 6 - 7 | 792 | 79 | |
BC006470.1 | - | 6 - 7 | 748 | 79 | |
BT006957.1 | - | 6 - 7 | 729 | 79 |