The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015201.4 | NP_056016.1 | 11 - 12 | 1492 | 134 |
GenBank mRNA | AB060694.1 | - | 10 - 11 | 1160 | 134 |
AK290677.1 | - | 11 - 12 | 1454 | 134 | |
AK293719.1 | - | 10 - 11 | 1136 | 134 | |
AK310015.1 | - | 9 - 10 | 1066 | 134 | |
AK315612.1 | - | 11 - 12 | 1461 | 134 | |
BC001086.2 | - | 10 - 11 | 1218 | 134 | |
BC005160.2 | - | 10 - 11 | 1245 | 134 | |
BC007274.1 | - | 11 - 12 | 1387 | 134 | |
BC013787.1 | - | 11 - 12 | 1454 | 134 | |
BC013980.2 | - | 11 - 12 | 1457 | 134 | |
BC017674.2 | - | 11 - 12 | 1444 | 134 | |
BC073813.1 | - | 6 - 7 | 751 | 134 | |
BC098446.1 | - | 7 - 8 | 888 | 134 | |
D50914.1 | - | 10 - 11 | 1232 | 134 |