The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130415.1 | NP_001123887.1 | 2 - 2 | 4633 | 112 |
NM_030817.2 | NP_110444.3 | 2 - 2 | 4541 | 112 | |
GenBank mRNA | AK024963.1 | - | 1 - 1 | 3427 | 112 |
AL136783.1 | - | 1 - 1 | 4467 | 112 |