The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001142434.1 | NP_001135906.1 | 9 - 10 | 2271 | 150 |
NM_012215.3 | NP_036347.1 | 10 - 11 | 2430 | 150 | |
XM_017015585.1 | XP_016871074.1 | 11 - 12 | 2807 | 150 | |
XM_017015586.1 | XP_016871075.1 | 7 - 8 | 1594 | 150 | |
GenBank mRNA | AB014579.2 | - | 10 - 11 | 1992 | 150 |
AF036144.2 | - | 10 - 11 | 2382 | 150 | |
AK002091.1 | - | 2 - 3 | 782 | 150 | |
BC001343.1 | - | 2 - 3 | 453 | 150 | |
BC039583.2 | - | 9 - 10 | 2230 | 150 |