The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001142434.1 | NP_001135906.1 | 13 - 14 | 2738 | 74 |
NM_012215.3 | NP_036347.1 | 14 - 15 | 2897 | 74 | |
XM_017015585.1 | XP_016871074.1 | 15 - 16 | 3274 | 74 | |
XM_017015586.1 | XP_016871075.1 | 11 - 12 | 2061 | 74 | |
GenBank mRNA | AB014579.2 | - | 14 - 15 | 2459 | 74 |
AF036144.2 | - | 14 - 15 | 2849 | 74 | |
AK002091.1 | - | 6 - 7 | 1249 | 74 | |
AK027008.1 | - | 2 - 3 | 254 | 74 | |
BC001343.1 | - | 6 - 7 | 920 | 74 | |
BC039583.2 | - | 13 - 14 | 2655 | 74 |