The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001142434.1 | NP_001135906.1 | 4 - 5 | 924 | 67 |
NM_012215.3 | NP_036347.1 | 4 - 5 | 924 | 67 | |
XM_017015585.1 | XP_016871074.1 | 5 - 6 | 1301 | 67 | |
GenBank mRNA | AB014579.2 | - | 4 - 5 | 486 | 67 |
AF036144.2 | - | 4 - 5 | 876 | 67 | |
AF307332.1 | - | 4 - 5 | 876 | 67 | |
BC039583.2 | - | 4 - 5 | 883 | 67 | |
BC047877.1 | - | 4 - 5 | 879 | 67 |