Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MYO1B
Entrez Gene ID
4430
Gene Name
myosin IB
Gene Aliases
myr1
Chromosome Location
Chr.2: 191245381 - 191425389 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130158.1 NP_001123630.1 13 - 14 1438 138
NM_001161819.1 NP_001155291.1 13 - 14 1276 138
NM_012223.3 NP_036355.2 13 - 14 1438 138
XM_005246572.1 XP_005246629.1 13 - 14 1302 138
XM_017004156.1 XP_016859645.1 13 - 14 1411 138
XM_017004157.1 XP_016859646.1 13 - 14 1411 138
XM_017004158.1 XP_016859647.1 11 - 12 1027 138
GenBank mRNA AB290176.1 - 13 - 14 1455 138
AK000160.1 - 9 - 10 772 138
BC053558.1 - 13 - 14 1276 138
BQ650834.1 - 3 - 4 264 138

View Full Product Details