The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130158.1 | NP_001123630.1 | 17 - 18 | 2034 | 75 |
NM_001161819.1 | NP_001155291.1 | 17 - 18 | 1872 | 75 | |
NM_012223.3 | NP_036355.2 | 17 - 18 | 2034 | 75 | |
XM_005246572.1 | XP_005246629.1 | 17 - 18 | 1898 | 75 | |
XM_017004156.1 | XP_016859645.1 | 17 - 18 | 2007 | 75 | |
XM_017004157.1 | XP_016859646.1 | 17 - 18 | 2007 | 75 | |
XM_017004158.1 | XP_016859647.1 | 15 - 16 | 1623 | 75 | |
GenBank mRNA | AB290176.1 | - | 17 - 18 | 2051 | 75 |
AK000160.1 | - | 13 - 14 | 1368 | 75 | |
BC053558.1 | - | 17 - 18 | 1872 | 75 |