The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130158.1 | NP_001123630.1 | 5 - 6 | 705 | 164 |
NM_001161819.1 | NP_001155291.1 | 5 - 6 | 543 | 164 | |
NM_012223.3 | NP_036355.2 | 5 - 6 | 705 | 164 | |
XM_005246572.1 | XP_005246629.1 | 5 - 6 | 569 | 164 | |
XM_017004156.1 | XP_016859645.1 | 5 - 6 | 678 | 164 | |
XM_017004157.1 | XP_016859646.1 | 5 - 6 | 678 | 164 | |
XM_017004158.1 | XP_016859647.1 | 3 - 4 | 294 | 164 | |
GenBank mRNA | AB290176.1 | - | 5 - 6 | 722 | 164 |
BC021665.1 | - | 5 - 6 | 853 | 164 | |
BC053558.1 | - | 5 - 6 | 543 | 164 | |
BG563224.1 | - | 5 - 6 | 495 | 164 |