The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014244.4 | NP_055059.2 | 4 - 5 | 996 | 62 |
NM_021599.3 | NP_067610.1 | 4 - 5 | 996 | 62 | |
XM_017010063.1 | XP_016865552.1 | 3 - 4 | 432 | 62 | |
GenBank mRNA | AJ003125.1 | - | 4 - 5 | 894 | 62 |
AK308213.1 | - | 4 - 5 | 999 | 62 |