The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001257230.1 | NP_001244159.1 | 24 - 25 | 2730 | 82 |
NM_001257234.1 | NP_001244163.1 | 24 - 25 | 2803 | 82 | |
NM_001257237.1 | NP_001244166.1 | 24 - 25 | 2907 | 82 | |
NM_001324292.1 | NP_001311221.1 | 24 - 25 | 2834 | 82 | |
NM_001324293.1 | NP_001311222.1 | 22 - 23 | 2733 | 82 | |
XM_011531030.1 | XP_011529332.1 | 25 - 26 | 3278 | 82 | |
XM_017029848.1 | XP_016885337.1 | 24 - 25 | 3182 | 82 | |
XM_017029855.1 | XP_016885344.1 | 17 - 18 | 2472 | 82 | |
GenBank mRNA | AK026671.1 | - | 9 - 10 | 963 | 82 |
AK223154.1 | - | 9 - 10 | 963 | 82 | |
AK300394.1 | - | 24 - 25 | 2672 | 82 | |
AK316306.1 | - | 24 - 25 | 2776 | 82 | |
AK316522.1 | - | 24 - 25 | 2851 | 82 | |
BC117377.1 | - | 9 - 10 | 961 | 82 | |
BC117379.1 | - | 9 - 10 | 961 | 82 |