Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CLN3
Entrez Gene ID
1201
Gene Name
CLN3, battenin
Gene Aliases
BTS, JNCL
Chromosome Location
Chr.16: 28466653 - 28492302 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000086.2 NP_000077.1 7 - 8 858 125
NM_001042432.1 NP_001035897.1 8 - 9 894 125
NM_001286104.1 NP_001273033.1 7 - 8 822 125
NM_001286105.1 NP_001273034.1 5 - 6 699 125
NM_001286109.1 NP_001273038.1 5 - 6 707 125
NM_001286110.1 NP_001273039.1 6 - 7 779 125
GenBank mRNA AF015593.1 - 7 - 8 589 125
AF015594.1 - 6 - 7 550 125
AF015596.1 - 4 - 5 356 125
AF077956.1 - 7 - 8 535 125
AF077957.1 - 7 - 8 535 125
AF077958.1 - 7 - 8 535 125
AF077959.1 - 7 - 8 535 125
AF077961.1 - 7 - 8 535 125
AF077962.1 - 7 - 8 535 125
AF077963.1 - 7 - 8 535 125
AF077966.1 - 7 - 8 535 125
AF077967.1 - 6 - 7 455 125
AF077971.1 - 7 - 8 535 125
AF077972.1 - 7 - 8 535 125
AF077973.1 - 5 - 6 376 125
AF078169.1 - 7 - 8 535 125
AK090709.1 - 9 - 10 1889 125
AK294070.1 - 6 - 7 703 125
AK294250.1 - 7 - 8 584 125
AK295500.1 - 7 - 8 782 125
AK297690.1 - 5 - 6 641 125
AK302027.1 - 7 - 8 562 125
AK302138.1 - 7 - 8 583 125
AK313002.1 - 8 - 9 666 125
BC002394.2 - 8 - 9 652 125
BC004433.1 - 8 - 9 652 125
BC111068.1 - 5 - 6 637 125
U32680.1 - 7 - 8 672 125

View Full Product Details