Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HORMAD2
Entrez Gene ID
150280
Gene Name
HORMA domain containing 2
Gene Aliases
CT46.2
Chromosome Location
Chr.22: 30080069 - 30207425 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011529914.2 XP_011528216.1 3 - 4 650 92
XM_011529917.2 XP_011528219.1 3 - 4 509 92
XM_011529918.1 XP_011528220.1 3 - 4 650 92
XM_017028621.1 XP_016884110.1 3 - 4 531 92
XM_017028622.1 XP_016884111.1 3 - 4 450 92
XM_017028624.1 XP_016884113.1 3 - 4 650 92
XM_017028625.1 XP_016884114.1 1 - 2 86 92
GenBank mRNA AK098703.1 - 3 - 4 475 92
BC030013.1 - 3 - 4 260 92
BI459310.1 - 3 - 4 255 92
CR456341.1 - 2 - 3 237 92
HY002382.1 - 4 - 5 300 92

View Full Product Details