The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_198566.3 | NP_940968.1 | 3 - 4 | 686 | 97 |
XM_011514038.2 | XP_011512340.1 | 3 - 4 | 595 | 97 | |
XM_017009445.1 | XP_016864934.1 | 2 - 3 | 717 | 97 | |
GenBank mRNA | AK056925.1 | - | 3 - 4 | 601 | 97 |
AK303336.1 | - | 2 - 3 | 362 | 97 | |
BC036867.1 | - | 3 - 4 | 549 | 97 |