The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_198321.3 | NP_938080.1 | 4 - 5 | 698 | 77 |
GenBank mRNA | AB078145.1 | - | 4 - 5 | 566 | 77 |
AJ505950.1 | - | 4 - 5 | 566 | 77 | |
AK074132.1 | - | 4 - 5 | 546 | 77 | |
AK127135.1 | - | 4 - 5 | 667 | 77 |