The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002581.4 | NP_002572.2 | 3 - 4 | 2006 | 96 |
XM_006717129.2 | XP_006717192.1 | 1 - 2 | 82 | 96 | |
XM_017014784.1 | XP_016870273.1 | 3 - 4 | 2046 | 96 | |
GenBank mRNA | BC078657.1 | - | 3 - 4 | 1636 | 96 |
X68280.1 | - | 3 - 4 | 1397 | 96 |