The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153445.1 | NP_703146.1 | 1 - 1 | 235 | 81 |
GenBank mRNA | AF158377.1 | - | 1 - 1 | 235 | 81 |