The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000749.3 | NP_000740.1 | 2 - 3 | 328 | 108 |
GenBank mRNA | BC069681.1 | - | 2 - 3 | 328 | 108 |
BC069703.1 | - | 2 - 3 | 328 | 108 | |
BC069788.1 | - | 2 - 3 | 328 | 108 | |
U62438.1 | - | 2 - 3 | 296 | 108 | |
X67513.1 | - | 1 - 2 | 95 | 108 | |
Y08417.1 | - | 2 - 3 | 328 | 108 |