Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DMXL2
Entrez Gene ID
23312
Gene Name
Dmx like 2
Gene Aliases
PEPNS, RC3
Chromosome Location
Chr.15: 51447724 - 51622833 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001174116.1 NP_001167587.1 2 - 3 439 79
NM_001174117.1 NP_001167588.1 2 - 3 439 79
NM_015263.3 NP_056078.2 2 - 3 439 79
XM_005254255.1 XP_005254312.1 2 - 3 502 79
XM_005254256.1 XP_005254313.1 2 - 3 502 79
XM_011521402.2 XP_011519704.1 2 - 3 502 79
XM_017022034.1 XP_016877523.1 2 - 3 175 79
GenBank mRNA AF389880.1 - 2 - 3 421 79
AK000156.1 - 2 - 3 440 79
BC140781.1 - 2 - 3 304 79
BC144539.1 - 2 - 3 304 79
BX491117.1 - 2 - 3 320 79
DA484900.1 - 2 - 3 502 79

View Full Product Details