Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MPV17L
Entrez Gene ID
255027
Gene Name
MPV17 mitochondrial inner membrane protein like
Gene Aliases
M-LPH, MLPH1, MLPH2, MPV17L1
Chromosome Location
Chr.16: 15395569 - 15409686 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173803.3 NP_776164.2 1 - 2 452 103
GenBank mRNA BC061514.1 - 1 - 2 401 103
DQ004256.1 - 1 - 2 452 103

View Full Product Details