The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004617.3 | NP_004608.1 | 3 - 4 | 691 | 82 |
GenBank mRNA | AK315464.1 | - | 3 - 4 | 578 | 82 |
BC001386.2 | - | 3 - 4 | 561 | 82 | |
BT007430.1 | - | 3 - 4 | 394 | 82 | |
U31449.1 | - | 3 - 4 | 559 | 82 |